VWR® Double-Sided Tape
# de catalogue 10853-860
Fournisseur: VWR International
Spécifications
- Length:36 yds.
- Width:1"
- Description:Lo Tack
- Cat. no.:10853-860
Spécifications
A propos de cet article
Transparent double coated polyester film available in Hi and Lo Tack with liner.
- Double-coated
- Polyester fabrication
Hi Tack features a permanent solvent acrylic adhesive—easily bonds dissimilar surfaces.
Lo Tack features a permanent adhesive on one side and removable on the other—ideal for peel and re-seal applications.
Customers Who Bought This Also Bought