ContactUsLinkComponent
VWR* Lo Tack Tape Double Coated
VWR® Double-Sided Tape
# de catalogue 10853-860
Fournisseur:  VWR International
VWR* Lo Tack Tape Double Coated
VWR® Double-Sided Tape
# de catalogue 10853-860
Fournisseur:  VWR International
Spécifications
  • Length:
    36 yds.
  • Width:
    1"
  • Description:
    Lo Tack
  • Cat. no.:
    10853-860
Spécifications
A propos de cet article
Transparent double coated polyester film available in Hi and Lo Tack with liner.
  • Double-coated
  • Polyester fabrication
Hi Tack features a permanent solvent acrylic adhesive—easily bonds dissimilar surfaces.
Lo Tack features a permanent adhesive on one side and removable on the other—ideal for peel and re-seal applications.
Customers Who Bought This Also Bought