Spécifications
- Sterility:Sterile
- Description:Standard Flock Tip, Polystyrene Handle with 80 mm Breakpoint
- Cat. no.:10852-164
- Cap Color:Red
- Supplier No.:25-3306-UBT
Spécifications
A propos de cet article
Get all your specimen collection devices in one place! Ideal for the collection and transport of specimen when no medium is required
Systems are designed for safe, sterile specimen transport. No matter which swab(s) are chosen, all transport systems include a round-bottom tube made from virgin polypropylene resin, which will securely hold the swab, maintaining a safe and sterile environment.
Tubes include a distinctive label describing product identity, expiry date, patient identification, and collection information. System applications include; specimen collection, rapid diagnostic testing, forensic evidence collection, DNA testing, and Buccal cell collection.
Choose from a range of PurFlock Ultra® styles and tip sizes. Single or double options are available.