Rtx®-5 GC Columns, Restek
Catalog # 10849-198
Supplier: Restek
Specifications
- Column Format:Capillary
- Film thickness:0.25 µm
- Ø int.:0.25 mm
- Description:Rtx-5
- Length:30 m
- Cat. no.:10849-198
- Supplier No.:10223-127
Specifications
About this item
A low-polarity general-purpose column for the analysis of pesticides, PCBs, aromatic hydrocarbons, drugs, and essential oils.
- Extremely low bleed
- Low-polarity column
- Internal dimensions and film thickness vary between models
Equivalent to USP G27, G36 phases. All residual catalysts and low molecular weight fragments are removed from the Rtx®-5 polymer, providing a tight mono-modal distribution and extremely low bleed. Phase composition: Crossbond® 5% diphenyl, 95% dimethylpolysiloxane.
Customers Who Bought This Also Bought