Order Entry
ContactUsLinkComponent
Wash Bottle 500mL
Nalgene® Wash Bottles, Right-to-Understand, Thermo Scientific
Catalog # 10799-049
Wash Bottle 500mL
Nalgene® Wash Bottles, Right-to-Understand, Thermo Scientific
Catalog # 10799-049

Specifications

  • Capacity:
    500 ml (16.9 oz.)
  • Description:
    Isopropanol
  • Cat. no.:
    10799-049
  • Cap Color:
    Yellow
  • Supplier No.:
    2428-0504

Specifications

About this item

Prevent chemical accidents in the lab with Right-to-Understand LDPE, translucent wash bottles.

  • Self-venting wash bottles eliminates the risk of bottles self-dispensing
  • Good chemical resistance
  • Good temperature resistance

Bottles are pre-labeled with chemical identification and safe handling information following the 100% Globally Harmonized System (GHS) specifications as applicable to secondary laboratory containers. Wash bottles are ready to use when you transfer common laboratory chemicals from containers.

Customers Who Bought This Also Bought