Order Entry
ContactUsLinkComponent
Laboratory Tubing 3/8in x 5/8in
Tubing, Vacuum, High Wall, Kimble Chase
# de catalogue 10847-220
Laboratory Tubing 3/8in x 5/8in
Tubing, Vacuum, High Wall, Kimble Chase
# de catalogue 10847-220

Spécifications

  • Length:
    10'
  • Cat. no.:
    10847-220
  • I.D.×O.D.:
    ⅜×⅝"
  • Supplier No.:
    869791-0038

Spécifications

A propos de cet article

The Tygon S3™ E-3603 laboratory tubing is used for drain, vent, and general laboratory applications, and is specially formulated for resistance to flex-fatigue and abrasion

  • Phthalate-free and BPA-free
  • Autoclavable and outstanding chemical resistance
  • Non-oxidizing and non-contaminating
  • Smooth inner bore helps prevent build
  • Vacuum rating 29.9" Hg
  • Coils marked at 1" intervals

It has heavy walls to withstand a full vacuum, and may be used on a variety of glassware including filter flasks, condensers, and distillation adapters.

Customers Who Bought This Also Bought