Respirator Filter Cartridges with Stacked P100 Filter 6000 Series, 3M™
# de catalogue CA56621-892
Fournisseur: 3M
Spécifications
- Cat. no.:CA56621-892
- Approved For Use Against:Certain organic vapors
- Cartridge Type:Organic Vapors/P100
- NIOSH Approval:TC-84A-0699
- Supplier No.:60921
Spécifications
A propos de cet article
For use with 3M™ 6000 series respirator facepieces.
- NIOSH-approved P100 filter is permanently attached
- Manufacturer’s time use restrictions of 40 hours of use or 30 days
- Certain Organic Vapors/Acid Gas/P100 approved for use against chlorine, hydrogen chloride, sulfur dioxide, hydrogen fluoride, hydrogen sulfide (escape only)
- Multi-Gas/Vapor/P100 approved for use against certain organic vapors, chlorine, chlorine dioxide, hydrogen chloride, ammonia, methylamine, sulfur dioxide, hydrogen fluoride, hydrogen sulfide (escape only)
NIOSH approved P100 filter is permanently attached. Manufacturer’s time use restrictions of 40 hours of use or 30 days, whichever is first, apply if used in environments containing only oil aerosols.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product. OSHA regulations require that gas-proof goggles be worn with half facepiece respirators when used against formaldehyde.
Customers Who Bought This Also Bought