Spécifications
- Injection Type:Plain preserved
- Cat. no.:470220-074
- Packaging:Vacuum pack of 10
- Preservative:Formalin preserved
Spécifications
A propos de cet article
Formalin preserved sheep eye. Available in jars or pails. Shipped in nontoxic Wardsafe.
- Economical
- High quality preservation
Available in plains or single injected. Single injected sheep eyes are injected with red latex to highlight the arterial circulation of the eye and its surrounding tissue.
Note: Ward's® preserved sheep materials is a byproduct of the food industry. Sheep are not raised specifically for dissection. These animals are recycled" for educational use."