ContactUsLinkComponent
DPBS (1X) W/OUT CA W/OUT MG 1L
VWR® Dulbecco's Phosphate Buffered Saline (DPBS)
Catalog # VWRL0119-1000
Supplier:  VWR
CAS Number:  
VWR® Dulbecco's Phosphate Buffered Saline (DPBS)
Catalog # VWRL0119-1000
Supplier:  VWR
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    DPBS (1X) w/out Ca, w/out Mg
  • Size:
    1000 ml
  • Cat. no.:
    VWRL0119-1000
Specifications
About this item
DPBS is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells.
  • Used to irrigate, wash, and dilute mammalian cells
  • Phosphate buffering maintains the pH in the physiological range
  • Available with or without calcium and magnesium
DPBS is used to irrigate, wash, and dilute mammalian cells. Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells.
Customers Who Bought This Also Bought