Specifications
- Description:DPBS (1X) w/out Ca, w/out Mg
- Size:500 ml
- Cat. no.:VWRL0119-0500
Specifications
About this item
DPBS is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells.
- Used to irrigate, wash, and dilute mammalian cells
- Phosphate buffering maintains the pH in the physiological range
- Available with or without calcium and magnesium
DPBS is used to irrigate, wash, and dilute mammalian cells. Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells.