Order Entry
ContactUsLinkComponent
Safety Can for Liquid Disposal
Liquid Disposal Safety Cans, Justrite®
# de catalogue 10836-904
Fournisseur:  Justrite
Safety Can for Liquid Disposal
Liquid Disposal Safety Cans, Justrite®
# de catalogue 10836-904
Fournisseur:  Justrite

Spécifications

  • Capacity:
    19 L (5 gal.)
  • Color:
    Red
  • Description:
    Safety can for liquid disposal
  • Cat. no.:
    10836-904
  • O.D.×H:
    305×508 mm
  • Supplier No.:
    14565

Spécifications

A propos de cet article

These high-density polyethylene (HDPE) disposal cans with built-in fill gauge provide superior chemical resistance and leak protection.

  • Superior chemical resistance and leak protection
  • Spring-loaded sealed cap automatically vents
  • Stainless steel flame arrester
  • Design resists accidental tip over

Built-in polyethylene fill gauge indicator pops up to warn you that the container is almost full.

Customers Who Bought This Also Bought