Specifications
- Description:AimTab™ Ketone Tablets
- Cat. no.:10837-000
- Supplier No.:13100
Specifications
About this item
AimTab™ Ketone Tablets are desgined for the semi-quantitative determination of ketones (acetoacetic acid and acetone) in urine, serum, plasma, or whole blood
- Specifically designed for acetoacetic acid and acetone
- Tablets will not react with betahydroxybutyric
- CLIA moderately complex when testing serum, plasma, or whole blood
- Same test procedure as Acetest®
Tablets detect as little as 5mg/dL of acetoacetic acid in urine.