ContactUsLinkComponent
AimTab* Ketones Tablets
AimTab™ Ketone Tablets, Germaine® Laboratories
Catalog # 10837-000
CAS Number:  
AimTab* Ketones Tablets
AimTab™ Ketone Tablets, Germaine® Laboratories
Catalog # 10837-000
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    AimTab™ Ketone Tablets
  • Cat. no.:
    10837-000
  • Supplier No.:
    13100
Specifications
About this item
AimTab™ Ketone Tablets are desgined for the semi-quantitative determination of ketones (acetoacetic acid and acetone) in urine, serum, plasma, or whole blood
  • Specifically designed for acetoacetic acid and acetone
  • Tablets will not react with betahydroxybutyric
  • CLIA moderately complex when testing serum, plasma, or whole blood
  • Same test procedure as Acetest®
Tablets detect as little as 5mg/dL of acetoacetic acid in urine.
Certifications: CLIA waived for urine.
Customers Who Bought This Also Bought