To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Description:AxyPrep™ Mag PCR Clean-Up Kit
- Size:227 Preps
- Description:AxyPrep™ Mag PCR Clean-Up Kit
- Cat. no.:97067-617
- Volume:5 ml
- Supplier No.:MAG-PCR-CL-5
Specifications
About this item
The efficient removal of unincorporated dNTPs, salts and enzymes after any PCR is detrimental to the success or failure of downstream applications, such as genotyping, Sanger, and Next Generation Sequencing. The AxyPrep™ Mag PCR Clean Up Kit utilizes a paramagnetic bead technology for rapid high throughput purification of PCR products. Additionally, the AxyPrep™ Mag PCR Clean Up Kit employs an optimized buffer to selectively bind PCR amplicons 60 bp and larger to paramagnetic beads, providing end users flexibility and high recovery options. The protocol comprises binding, washing and eluting steps, where primers, nucleotides, salts and enzymes in the reaction mixture are efficiently removed leaving a purified PCR product that is essentially free of contaminants. No centrifugation or filtration steps are required, allowing the process to be adapted to multiple commercially available automation platforms.
- Flexible clean up: High recovery of amplicons with the option to choose recovery of >60 or >100 bp
- Efficient removal of unincorporated dNTPs, primers, primer dimers, salts and other contaminants
- Manual and automation friendly protocol
- Processing time: 15 minutes/96 samples
- No centrifugation or filtration required