ContactUsLinkComponent
Loading dye base 50X
VWR® Loading Dye Base 50X
# de catalogue 10836-726
Fournisseur:  VWR
Numéro CAS:  
VWR® Loading Dye Base 50X
# de catalogue 10836-726
Fournisseur:  VWR
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Loading Dye Base 50X
  • Size:
    25 mL
  • Supplier No.:
    1B1812-25ML
Spécifications
A propos de cet article
Loading Dye Base, 50X is used for custom loading buffer preparation.
  • Custom loading buffer preparation
  • Concentrated solution containing patent blue
  • Eliminates the need to weigh messy dye powders
Loading Dye Base, 50X is used for custom loading buffer preparation. The Loading Dye Base is a concentrated solution containing patent blue, bromophenol blue and amaranth tracking dyes, which migrate at approximately 4,000 bp, 400 bp and 10 bp, on a 1% agarose gel, respectively. Using Loading Dye Base eliminates the need to weigh messy dye powders.
Customers Who Bought This Also Bought