Alligator clip wires
# de catalogue 470219-722
Fournisseur: United Scientific Supplies
Spécifications
- Length:24"
- Description:Alligator clip wire pack of 6
- Cat. no.:470219-722
- Supplier No.:WAG024-PK/6
Spécifications
A propos de cet article
Alligator clip wires can connect to items that don't have banana plugs.
- Protective Vinyl Sheath
- Various Lengths and Pack Sizes Available
- Flexible, 18-Gauge Stranded Wire Cord
A flexible, 18-gauge stranded wire cord has alligator clips at both ends.
Alligator clips have a protective vinyl sheath. They're available in various lengths and pack sizes.
Customers Who Bought This Also Bought