VWR® Qualitative Fluted Circles, Folded, Grade 313
# de catalogue CA28333-065
Fournisseur: VWR International
Spécifications
- Ø:18.5 cm
- Particle retention:5 µm
- Flow rate:Medium
- Porosity:Medium fine
Spécifications
A propos de cet article
Used for drugs, pharmaceuticals, liquors, tinctures, chromatography, and protein-free filtrations in the clinical laboratory.
- Medium fine porosity
- Medium flow rate
- Assorted sizes
Fluted circles are available in a variety of sizes and boast a medium flow rate.
Customers Who Bought This Also Bought