Order Entry
ContactUsLinkComponent
Cap, 12mm solid top storage, 13-425 Closure screw Thread, Color: Black, Material: Polypropylene
Caps for 12-425 Screw-Thread Vials
# de catalogue 10820-602
Fournisseur:  Thermo Scientific
Caps for 12-425 Screw-Thread Vials
# de catalogue 10820-602
Fournisseur:  Thermo Scientific

Spécifications

  • Cap Size:
    12-425
  • Cap Type:
    Screw cap
  • Ø:
    12 mm
  • Septum:
    None
  • Description:
    Solid top cap
  • Material:
    PP
  • Packaging:
    500/pack
  • For Use With:
    12-425 screw thread vials
  • Cap Color:
    Black
  • Supplier No.:
    12-SCS

Spécifications

A propos de cet article

Screw thread caps feature a 13 to 425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs.

  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Phenolic resin caps perform well at high temperatures and are compatible with exposure to corrosives
  • Caps are bonded septa resist dislodging during injection when using large diameter blunt needles

Convenient pre-assembled caps and septa minimize contamination from handling.

Customers Who Bought This Also Bought