ContactUsLinkComponent
Bath Brite
Bath Brite, FoxWest Sales
# de catalogue 10820-596
Fournisseur:  Decon Labs
Bath Brite
Bath Brite, FoxWest Sales
# de catalogue 10820-596
Fournisseur:  Decon Labs
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Bath Brite
  • Cat. no.:
    10820-596
  • Supplier No.:
    3408
Spécifications
A propos de cet article
Bath Brite is designed for use in water baths, incubators, circulators, and other similar temperature controlled lab equipment.
Adding 10 to 15 drops of Bath Brite per gallon of water will keep water clear and fresh, and extend the time between water replacement. Supplied in a 237 mL (8 oz.) bottle with a flip-top cap for easy dispensing.
Customers Who Bought This Also Bought