Spécifications
- Description:Bath Brite
- Cat. no.:10820-596
- Supplier No.:3408
Spécifications
A propos de cet article
Bath Brite is designed for use in water baths, incubators, circulators, and other similar temperature controlled lab equipment.
Adding 10 to 15 drops of Bath Brite per gallon of water will keep water clear and fresh, and extend the time between water replacement. Supplied in a 237 mL (8 oz.) bottle with a flip-top cap for easy dispensing.