ContactUsLinkComponent
Ampule
Spore Ampoules, Biological Indicator, Namsa
Catalog # 10820-626
Supplier:  NAMSA
CAS Number:  
Spore Ampoules, Biological Indicator, Namsa
Catalog # 10820-626
Supplier:  NAMSA
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Pack type:
    10/Box
  • Description:
    Negative Control
  • Cat. no.:
    10820-626
  • Volume:
    1 ml
Specifications
About this item
Spore Ampoules are ideal for use in validation and routine monitoring of liquid steam sterilization cycles
  • Reduced incubation time of 48 hours
  • Each snap top ampoule is imprinted with lot number
  • Saves time by eliminating aseptic culturing required with traditional spore strips
Unique packaging insert keeps ampoules in upright position and allows for quick reference to the number of units remain.
They are manufactured using hermetically sealed Type I borosilicate glass containing an exclusively formulated Tryptic Soy Broth (TSB) modified with pH indicator, and Geobacillus stearothermophilus spores. It shows clear visible results as growth is evident by turbidity and/or a color shift of the media from purple to yellow.
Certifications: Compliant with ISO 11138-1 and 11138-3
Delivery: Each pack is available with certificate of analysis and instructions for use.
Caution: Note: Storage under refrigerated conditions (2°C to 8°C) is required. Do not freeze spore ampoules.
Customers Who Bought This Also Bought