Sodium chloride, VWR® USP, Ph. Eur., BP, JP endotoxin tested
# de catalogue VWRB241-1KG
Fournisseur: Avantor Performance Materials
Numéro CAS: 7647-14-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:HDPE bottle
- Size:1 kg
- Cat. no.:VWRB241-1KG
- Supplier No.:VWRB241-1KG
Spécifications
Résultats des tests de spécification
- Acidity or Alkalinity:Passes Test
- Aluminum:≤ 0.2 ppm
- Appearance of Solution:Passes Test
- Arsenic:≤ 1.0 ppm
- Endotoxin:≤ 2.5EU/g
- Assay (dried basis):99.0% - 105%
- Barium Test:Passes Test
- Ferrocyanides:Passes Test
- Heavy Metals:≤ 3.0 ppm
- Identification A:Passes Test
- Identification B:Passes Test
- Iodides:Passes Test
- Iron:≤ 2.0 ppm
- Limit of Bromides:≤ 100 ppm
- Limit of Phosphates:≤ 25 ppm
- Limit of Potassium:≤ 500 ppm
- Loss on Drying:≤ 0.5%
- Magnesium and Alkaline - Earth Metals:≤ 100 ppm
- Nitrites:Passes Test
- Sulfate:≤ 200 ppm
- Sodium chloride, VWR® USP, Ph. Eur., BP, JP endotoxin tested:
Résultats des tests de spécification
A propos de cet article
Synonymes: Saline,Sea salt
Formule:NaCl
Point d'ébullition:1413 °C (1013 hPa)
Point de Fusion:801 °C
Densité:2,16 g/cm³ (20 °C)
Numéro MDL:MFCD00003477
Numéro CAS:7647-14-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 1413 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: 801 °C
:Pt Eclair:|Point eclaire:
:M.W. N/A N/A
Index Merck:14,08599
:Sodium chlorure
EINECS:231-598-3
:801
:1413
Température d'entreposage:Température ambiante
:12,08742
:Sodium chloride
:Storage Temperature: Ambient
Point d'ébullition:1413 °C (1013 hPa)
Densité:2.16 g/cm³ (20 °C)
:Length
:Width
:Height
:Diameter
:Thickness
:Boiling Pt:
:Boiling Pt: 1413 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 801 °C
:Flash Pt:
:Flash Pt:
:M.W. 58.44 g/mol
Température d'entreposage:Ambient
Customers Who Bought This Also Bought