ContactUsLinkComponent
Manometer
Manometer Demonstration
# de catalogue 470219-420
Fournisseur:  Wards
Manometer
Manometer Demonstration
# de catalogue 470219-420
Fournisseur:  Wards
Spécifications
  • Description:
    Manometer Demonstration
  • Cat. no.:
    470219-420
  • Supplier No.:
    PH0203G
Spécifications
A propos de cet article
Demonstrate the effect of pressure to water depth.
  • Complete package to highlight the variation of pressure in a liquid with depth
It consists of a pressure capsule of 35mm diameter placed in a bent steel tube. The pressure exerted by the water on the membrane of the capsule is measured by a pressure gauge mounted on a graduated support, connected to the probe by means of a transparent flexible tube. A device used to turn the capsule without leaving the water. The manometer tube is slideable on the support to facilitate setting of zero. Supplied with beaker.
Customers Who Bought This Also Bought