Order Entry
ContactUsLinkComponent
Dye
Tissue Marking Dye, Flip-Top, Cancer Diagnostics
# de catalogue 10809-812
Numéro CAS:  
Dye
Tissue Marking Dye, Flip-Top, Cancer Diagnostics
# de catalogue 10809-812
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Color:
    Black
  • Description:
    Tissue Marking Dye
  • Size:
    0.5 oz.
  • Cat. no.:
    10809-812
  • Supplier No.:
    0723-1

Spécifications

A propos de cet article

Tissue marking dyes are used for marking margins & multiple biopsies.

  • Margins defined without bleeding
  • Dyes won't change color or fade
  • Flip-top dispenser cap

Tissue marking dyes for marking margins & multiple biopsies. Dyes have been developed to define margins without bleeding, changing color or fading. Flip-top dispenser cap.

Customers Who Bought This Also Bought