ContactUsLinkComponent
Tube
VACUETTE® Urine Tubes, Greiner Bio-One
# de catalogue 10809-036
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
Tube
VACUETTE® Urine Tubes, Greiner Bio-One
# de catalogue 10809-036
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Capacity:
    10 ml
  • Ø:
    16 mm
  • Description:
    Tubes, Round-Bottom, with Non-Ridged Pull Cap
  • Height:
    100 mm
Spécifications
A propos de cet article
Urine tubes are designed for use as collection, transportation, or analysis vessels.
VACUETTE® urine tubes are also available with or without a stabilizing preservative. Tubes containing a preservative are intended for use with specimens that cannot be analyzed within two hours of collection, specimens being tested for an otherwise unstable analyte, or specimens being stabilized for microbiological studies. Tubes with preservative should be inverted 6–8 times following collection in order to acheive homogenous mixing.
  • Clear, durable plastic construction
  • Sterile
  • Leakproof and unbreakable
  • Disposable
Urine tubes are designed for use as collection, transportation, or analysis vessels.
Manufactured from clear, disposable plastic, these tubes are sterile, leak-proof, and virtually unbreakable
VACUETTE® urine tubes are also available with or without a stabilizing preservative. Tubes containing a preservative are intended for use with specimens that cannot be analyzed within two hours of collection, specimens being tested for an otherwise unstable analyte, or specimens being stabilized for microbiological studies. Tubes with preservative should be inverted 6–8 times following collection in order to acheive homogenous mixing.
Available in round or conical bottom formats. Round bottom tubes are used for chemical urine tests. Conical bottom tubes are used for microscopic examinations of urine sedimentation. All tubes are supplied with yellow caps.
Customers Who Bought This Also Bought