VWR® Spatulas, PTFE-Coated
# de catalogue 10806-410
Fournisseur: VWR International
Spécifications
- Length:—
- Description:VWR® Hayman spatula
- Cat. no.:10806-410
Spécifications
A propos de cet article
VWR® PTFE-Coated Spatulas are available in a variety of styles to accommodate an array of laboratory tasks.
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
All spatulas feature stainless steel construction, with a PTFE coating.
Customers Who Bought This Also Bought