Order Entry
ContactUsLinkComponent
VWR* Ear Punch, scissor style, Length: 2mm
VWR® Animal Ear Punches
# de catalogue 10806-294
Fournisseur:  VWR International
VWR* Ear Punch, scissor style, Length: 2mm
VWR® Animal Ear Punches
# de catalogue 10806-294
Fournisseur:  VWR International

Spécifications

  • Ø:
    2 mm Hole Size
  • Length:
    10.8 cm (4.25")
  • Description:
    Animal Ear Punch, Scissor-Style
  • Material:
    Surgical Grade Stainless Steel
  • Cat. no.:
    10806-294

Spécifications

A propos de cet article

VWR® Animal Ear Punches are made from corrosion-resistant ASTM (899-94) Series 400 stainless steel.

  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods

These ear punches are available in multiple designs, including thumb-style, plier-style, or scissor-style punches. Both the plier-style and scissor-style punches feature a longer handle for improved grip and handling.

Customers Who Bought This Also Bought