Formaldehyde Solution Spill Kit
Catalog # 470219-190
Supplier: Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Formaldehyde Solution Spill Kit
- Cat. no.:470219-190
- Supplier No.:IS5035
Specifications
About this item
Be prepared for spills in your lab.
- Enough Material to Clean One Liter of a 10% Formalin Solution
- Instructions and SDS
- Vinyl Exam Gloves
This kit includes a dust pan and brush to properly dispose of spills.
It also includes vinyl exam gloves, so you are protected while cleaning.
Ordering information: This kit includes an instruction sheet/SDS, vinyl exam gloves, polybags, twist ties, blank shipping tags, dust pan and brush, vermiculite, and an absorbent formaldehyde neutralizing agent (sodium bisulfite).
Customers Who Bought This Also Bought