ContactUsLinkComponent
Cap, Autosampler, screw top, with fitted PTFE septa, For use with 9-425mm, 12x32mm vials
Screw Caps, 9 mm MicroSolv™
# de catalogue 10804-662
Cap, Autosampler, screw top, with fitted PTFE septa, For use with 9-425mm, 12x32mm vials
Screw Caps, 9 mm MicroSolv™
# de catalogue 10804-662
Spécifications
  • Cap Size:
    9-425 mm
  • Cap Type:
    Screw cap
  • Septum:
    PTFE
  • Material:
    PP
  • Cat. no.:
    10804-662
  • Cap Color:
    Clear
  • Supplier No.:
    9502S-50C-N