Vial
# de catalogue 10804-190
Fournisseur: MicroSolv Technology Corp
Customers Who Bought This Also Bought
Potassium dihydrogen phosphate ≥99.0% ACS, VWR Chemicals BDH®
Fournisseur: VWR International
143,96 $ to 1 334,21 $
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Fournisseur: Ricca Chemical
175,32 $ to 1 620,42 $
Sodium chloride ≥99.0% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
116,87 $ to 467,59 $
Hydrofluoric acid 47 - 51%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Fournisseur: BDH
682,08 $