Screw Caps, MicroSolv™
# de catalogue 10804-132
Fournisseur: MicroSolv Technology Corp
Spécifications
- Cap Size:13 - 425 mm
- Cap Type:Screw cap
- Septum:Silicone Rubber / PTFE Pre-Slit
- Material:PP
- Cat. no.:10804-132
- Cap Color:White
- Supplier No.:9504S-30C-W
Spécifications
A propos de cet article
These screw caps are designed for use with 13-425 cap size, 15x45mm vials.
Customers Who Bought This Also Bought