Ferrule, HPLC, Endure brand ETFE. 1/4-28 thread male nuts and 1/8 In tubing sold separately
# de catalogue 10803-852
Fournisseur: MicroSolv Technology Corp
Ferrule, HPLC, Endure brand ETFE. 1/4-28 thread male nuts and 1/8 In tubing sold separately
# de catalogue 10803-852
Fournisseur: MicroSolv Technology Corp
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$45.27
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Fournisseur: MicroSolv Technology Corp
$438.81
WHEATON® MicroLiter Snap-Top Vials, 11 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$106.31
SureSTART™ Glass Crimp Top Vials, 2 ml, evel 1 Everyday Analysis, Thermo Scientific
Fournisseur: Thermo Scientific
$283.77 to $283.77
WHEATON® MicroLiter 9 mm Screw-Thread Vials, Component Kits, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$167.84
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$99.27 to $725.06
WHEATON® MicroLiter Crimp-Top Vials, 11 mm,Component Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$79.72