ContactUsLinkComponent
Disposable tips, Liquid handling 200Ul Conductive filtered 17280 Pce
Conductive Liquid Handling (LiHa) Disposable Tips, Tecan
# de catalogue CA10024-164
Fournisseur:  TECAN US INC. CA
Conductive Liquid Handling (LiHa) Disposable Tips, Tecan
# de catalogue CA10024-164
Fournisseur:  TECAN US INC. CA
Spécifications
  • Volume:
    200 µl
  • Filter:
    +
  • Tip Style:
    Tecan standard disposable tips, filtered
  • Supplier No.:
    10612553
Spécifications
A propos de cet article
These disposable tips are carefully designed, produced and validated to ensure reliable performance using Liquid Handling (LiHa) Arms and Flexible Channel (FCA) arms on Tecan platforms.
  • Available in a variety of tip volumes, with or without filters
  • Made of conductive polypropylene
  • Allow capacitive liquid level detection
  • Full product traceability
Tecan standard purity tips are supplied in space-saving cardboard packages and are used in applications such as ELISA, sample handling and biochemical screening.
Tecan pure purity tips are packaged in blister packs, and each lot of tips is tested and certified free from human DNA, DNase, RNase and PCR inhibitors by an external accredited laboratory and are used in applications such as genomics research, forensics and molecular diagnostics.
Tecan sterile purity tips are tested on a lot-by-lot basis in an external accredited laboratory, and are free from human DNA, DNase, RNase and endotoxins. These are certified to be sterile, with a guaranteed Sterility Assurance Level (SAL) of 10⁻⁶ and are supplied packaged in ANSI/SLAS-format boxes and are used in applications such as cell biology, microbiology and cell-based screening.
Delivery: Each tray of Tecan disposable tips is supplied with a Certificate of Conformity (CoC).
Customers Who Bought This Also Bought