Thermometer
# de catalogue 10024-982
Fournisseur: Thermco
Customers Who Bought This Also Bought
Water, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
$277.09 to $740.95
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
$635.20 to $2,541.50
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Fournisseur: VWR International
$66.85 to $318.03
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$194.14 to $694.12
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
$377.44 to $992.90
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
$500.19 to $1,790.36