Spécifications
- Color:Blue
- Description:Foam Biopsy Pads
- Cat. no.:10022-166
- Supplier No.:190195
Spécifications
A propos de cet article
The Globe Scientific foam biopsy pads are used to hold biopsies in place and prevent their loss during processing
They are produced from a special polyester urethane foam, which is verified for consistency throughout in order to ensure optimum solvent flow. Simply sandwich the biopsy sample between two pads and insert into a cassette for processing.