Dispensette® S Bottle Top Dispensers
# de catalogue 10018-948
Fournisseur: Brandtech
Spécifications
- Accuracy:≤±0.5%
- Volume:0.2 - 2 ml
- Description:Analog volume adjustment, with recirculation valve
- Environmentally Preferable:Sustainable Packaging
- Cat. no.:10018-948
- Precision:≤±0.1%
- Supplier No.:4600121
Spécifications
A propos de cet article
Fixed volume or variable volume, analogue-adjustable or digital easy calibration.
- With or without recirculation valve
- Autoclavable at 121 °C
- Three different models of volume adjustment available: Fixed (fixed volume for standard applications; simple calibration adjustment with supplied tool) and Analogue (analogue volume display, fast volume adjustment; simple calibration adjustment with supplied tool)
- Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy calibration technique for adjustment in seconds without tools; alteration of factory setting is automatically indicated
- Easy to calibrate and adjust in order to comply with ISO 9001 and GLP guidelines; alteration of factory setting is automatically indicated
Dispensette® S supports a wide range of applications for the dispensing of aggressive reagents directly from the supply bottle; such as concentrated bases and acids like H₃PO₄, H₂SO₄ (with certain exceptions such as HCl, HNO₃, HF, and so on), saline solutions, and a variety of organic solvents. Parts that are in contact with the liquid are made from borosilicate glass, ceramic, platinum-iridium, ETFE, FEP, PFA, PTFE and PP (discharge tube safety screw cap). Colour code: Red.
* Special fixed volumes: 0.5 to 100 ml (please state when ordering)!
Ordering information: Dispensette® S is supplied with a DE-M marking, performance certificate, telescopic filling tube, recirculation tube (optional), mounting tool and PP adapters.
Customers Who Bought This Also Bought