ContactUsLinkComponent
Standard
Orion™ Conductivity Standards, Solutions, and Accessories, Thermo Scientific
# de catalogue 52458-149
Numéro CAS:  
Standard
Orion™ Conductivity Standards, Solutions, and Accessories, Thermo Scientific
# de catalogue 52458-149
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    100 µS/cm Conductivity Standard, 47 ppm as NaCl TDS Standard
  • Size:
    5×60 ml
  • Supplier No.:
    011008
Spécifications
A propos de cet article
Perform virtually any electrochemistry analysis with Orion™ Conductivity Standards and Solutions
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
Conductivity standard temperature tables are programmed into Orion™ meter software for use with the automatic calibration feature.
Choose from a complete selection of conductivity accessories including conductivity standards and TDS (total dissolved solids) standards. All conductivity standards are traceable to NIST standard reference materials.
Customers Who Bought This Also Bought