Spécifications
- Description:100 µS/cm Conductivity Standard, 47 ppm as NaCl TDS Standard
- Size:5×60 ml
- Supplier No.:011008
Spécifications
A propos de cet article
Perform virtually any electrochemistry analysis with Orion™ Conductivity Standards and Solutions
- Traceable to NIST standard reference materials
- Designed to meet all end user requirements, including laboratory grade and DIN standards
Conductivity standard temperature tables are programmed into Orion™ meter software for use with the automatic calibration feature.
Choose from a complete selection of conductivity accessories including conductivity standards and TDS (total dissolved solids) standards. All conductivity standards are traceable to NIST standard reference materials.