ContactUsLinkComponent
Shoe Cover
DuPont™ Tyvek® 400 Shoe Covers
Catalog # 10016-952
Supplier:  DuPont
Shoe Cover
DuPont™ Tyvek® 400 Shoe Covers
Catalog # 10016-952
Supplier:  DuPont
Specifications
  • Garment application:
    Chemical protection
  • Color:
    White
  • Height:
    8 1/4"
  • Cat. no.:
    10016-952
  • Supplier No.:
    TY450SWH00LG
Specifications
About this item
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Two shoe cover height options available
  • Serged seams
  • One size fits most
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include: lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Certifications: Thickness meets ASTM D1777.
Customers Who Bought This Also Bought