DuPont™ Tyvek® 400 Shoe Covers
Catalog # 10016-952
Supplier: DuPont
Specifications
- Garment application:Chemical protection
- Color:White
- Height:8 1/4"
- Cat. no.:10016-952
- Supplier No.:TY450SWH00LG
Specifications
About this item
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
- Two shoe cover height options available
- Serged seams
- One size fits most
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include: lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Certifications: Thickness meets ASTM D1777.
Customers Who Bought This Also Bought