DuPont™ ProShield® 10 Coveralls with Standard Hood
Catalog # 10016-262
Supplier: DuPont
Specifications
- Clothing Size:XXL
- Garment application:Industrial,Laboratory
- Color:Blue
- Cat. no.:10016-262
- Chest Size:28 3/4"
- Inseam:28 1/4"
- Supplier No.:PB127SBU2X00
- Sleeve:36 1/4"
Specifications
About this item
DuPont™ ProShield® 10, spun bond melt blown spun bond (SMS) garments, are made to the high-quality standards you have come to expect from DuPont.
- Standard fit hood
- Elastic wrists and ankles
- Elastic waist
- Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments.
DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Garments are available in a variety of coverall styles in a choice of white, blue and gray. Applications include: general maintenance, janitorial/cleaning and other dirty work.
Attached hood features elastic around the face opening. Elastic openings and wrists and ankles allows for a tighter fit.
Certifications: Thickness meets ASTM D1777.
Customers Who Bought This Also Bought