Spécifications
- Description:Low-Profile 819 Disposable Blades
- Cat. no.:10015-014
- Supplier No.:14035838925
Spécifications
A propos de cet article
Disposable low-profile and high-profile microtome blades for routine sectioning.
- Stainless steel construction
- Ultra-fine blade
- Durable
- Coating provides higher-quality sectioning ribbons
Low-Profile 819 Disposable Blades are 80mm in length, 8mm high, and 0.25mm thick
Made of stainless steel, these blades are ultra-fine yet durable. They can be used with the Leica microtome and cryostat series, as well as for common cryostat and microtome models. An inventive blade coating process provides perceptibly improved results in section stretching and in the quality of sectioning ribbons in routine histology applications. High-Profile 818 Disposable Blades are made of stainless steel and offer exceptional durability. Ultra-fine yet durable, they are 80mm in length, 14mm high, and 0.35mm thick. These blades are suitable for use with the Leica microtomes and cryostat series, as well as for common cryostat and microtome models. An inventive blade coating process provides perceptibly improved results in section stretching and in the quality of sectioning ribbons in routine histology.
Premium Low-Profile Surgipath® DB80 LS blades cut the very thin sections needed for diagnostic clarity. The DB80 LS blades provide a new level of quality with a low-profile shape, a premium finish, and edge-to-edge, blade-to-blade consistency.
Premium High-Profile Disposable Surgipath® DB80 HS blades are ideal for thicker sections. Use the Surgipath® DB80 HS blades for a high-profile microtome and cryostat applications, as they offer sharpness, longevity, and consistency.
The High-Profile Disposable Microtome blade is a unique blade that rivals the traditional reusable microtome blade for routine sectioning. It offers heavier gauge steel than most disposable brands, eliminating any vibrating issues when cutting through extremely hard or fibrous specimens.