ContactUsLinkComponent
Blade
Surgipath® Sectioning Blades, Leica
# de catalogue 10015-014
Fournisseur:  Leica Biosystems
Blade
Surgipath® Sectioning Blades, Leica
# de catalogue 10015-014
Fournisseur:  Leica Biosystems
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Low-Profile 819 Disposable Blades
  • Cat. no.:
    10015-014
  • Supplier No.:
    14035838925
Spécifications
A propos de cet article
Disposable low-profile and high-profile microtome blades for routine sectioning.
  • Stainless steel construction
  • Ultra-fine blade
  • Durable
  • Coating provides higher-quality sectioning ribbons
Low-Profile 819 Disposable Blades are 80mm in length, 8mm high, and 0.25mm thick
Made of stainless steel, these blades are ultra-fine yet durable. They can be used with the Leica microtome and cryostat series, as well as for common cryostat and microtome models. An inventive blade coating process provides perceptibly improved results in section stretching and in the quality of sectioning ribbons in routine histology applications. High-Profile 818 Disposable Blades are made of stainless steel and offer exceptional durability. Ultra-fine yet durable, they are 80mm in length, 14mm high, and 0.35mm thick. These blades are suitable for use with the Leica microtomes and cryostat series, as well as for common cryostat and microtome models. An inventive blade coating process provides perceptibly improved results in section stretching and in the quality of sectioning ribbons in routine histology.
Premium Low-Profile Surgipath® DB80 LS blades cut the very thin sections needed for diagnostic clarity. The DB80 LS blades provide a new level of quality with a low-profile shape, a premium finish, and edge-to-edge, blade-to-blade consistency.
Premium High-Profile Disposable Surgipath® DB80 HS blades are ideal for thicker sections. Use the Surgipath® DB80 HS blades for a high-profile microtome and cryostat applications, as they offer sharpness, longevity, and consistency.
The High-Profile Disposable Microtome blade is a unique blade that rivals the traditional reusable microtome blade for routine sectioning. It offers heavier gauge steel than most disposable brands, eliminating any vibrating issues when cutting through extremely hard or fibrous specimens.
Customers Who Bought This Also Bought