ContactUsLinkComponent
FastSCRIPT* cDNA Synthesis kit
FastSCRIPT™ cDNA Synthesis Kit, Tonbo Biosciences
# de catalogue 10002-278
Fournisseur:  Tonbo Biosciences
Numéro CAS:  
FastSCRIPT* cDNA Synthesis kit
FastSCRIPT™ cDNA Synthesis Kit, Tonbo Biosciences
# de catalogue 10002-278
Fournisseur:  Tonbo Biosciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    FastSCRIPT™ cDNA Synthesis Kit
  • Cat. no.:
    10002-278
  • No. of Reactions:
    25
  • Supplier No.:
    31-5300-0025R
Spécifications
A propos de cet article
The FastSCRIPT cDNA synthesis kit contains everything to generate unbiased, efficient, sensitive high-quality cDNA for downstream experiments
The kit contains a proprietary M-MLV reverse transcriptase (RT) molecule blended with an optimized RNase inhibitor, a mix that is both thermostable and extremely active.
This RTase mix synthesizes cDNA at a temperature range of 45–55°C, providing increased specificity, higher yields of cDNA, and more full-length product than other reverse transcriptases. The relative concentrations of random hexamers and anchored oligo(dT) primers have been designed specifically to generate cDNA optimized for use in real-time PCR experiments.
The FastSCRIPT cDNA synthesis kit can be used with total or poly(A)+ RNA, miRNA and viral RNA targets and provides sensitive detection of low copy number transcripts.
and removes the need for user optimization, due to its robust performance and ease of use.
Customers Who Bought This Also Bought