ContactUsLinkComponent
Safety goggle, For use with UV light or Blue light, Hold down clamp, Use with RIGID* Miter Gauge
UV Safety Goggles, Azure Biosystems
# de catalogue 10001-820
Fournisseur:  Azure Biosystems
Product available on GSA Advantage®
UV Safety Goggles, Azure Biosystems
# de catalogue 10001-820
Fournisseur:  Azure Biosystems
Product available on GSA Advantage®
Spécifications
  • Description:
    UV safety goggles
  • Supplier No.:
    AC1022
Spécifications
A propos de cet article
Safety goggle used while working with UV light or blue light.
Recommended to be used for band excision.
Customers Who Bought This Also Bought