UV Safety Goggles, Azure Biosystems
# de catalogue 10001-820
Fournisseur: Azure Biosystems
Product available on GSA Advantage®
Spécifications
- Description:UV safety goggles
- Supplier No.:AC1022
Spécifications
A propos de cet article
Safety goggle used while working with UV light or blue light.
Recommended to be used for band excision.
Customers Who Bought This Also Bought