Order Entry
ContactUsLinkComponent
Caspase-3 Substrate
NucView® Caspase-3 Enzyme Substrates, Biotium
# de catalogue 10001-596
Fournisseur:  Biotium
Numéro CAS:  
Caspase-3 Substrate
NucView® Caspase-3 Enzyme Substrates, Biotium
# de catalogue 10001-596
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    NucView® 530 Caspase-3/7 enzyme substrate, 1 mM in DMSO
  • Size:
    100 µl
  • Cat. no.:
    10001-596
  • Supplier No.:
    10406

Spécifications

A propos de cet article

NucView® Caspase-3 Substrates are novel fluorescent probes that allow real-time detection of apoptosis by monitoring caspase-3/7 activity in intact cells using confocal microscopy, flow cytometry or microplate readers.

  • Real-time apoptosis monitoring: non-interfering with caspase activity unlike other caspase substrates or inhibitor-based assays
  • Bi-functional substrates: simultaneous detection of caspase 3/7 activity and visualization apoptotic nuclear morphology
  • Rapid, no-wash assay for microscopy or flow cytometry
  • Stable for several days for performing time-course assays
  • Tolerant of formaldehyde fixation and permeabilization
  • For use in adherent or suspension cell cultures
  • Storage at 4 °C

NucView® substrates consist of a fluorogenic DNA dye coupled to the caspase-3/7 DEVD recognition sequence. Upon entering apoptotic cells, the non-fluorescent substrate is cleaved by caspases-3/7 releasing a high-affinity DNA binding dye which migrates to the nucleus and becomes fluorescent upon binding to DNA.

NucView® 488 Caspase-3 Substrate, is also offered as a 1 mM solution in PBS (phosphate-buffered saline) for use in cells that are sensitive to DMSO toxicity. NucView® 405 and 530 are also available in 1 mM in PBS and trial sizes are available for both PBS and DMSO formulations. Note: In non-DMSO sensitive cell types, adding DMSO during the substrate incubation may enhance NucView staining.

Available in 3 color options:
NucView® 488: Ex/Em (bound to DNA) 500 nm/530 nm, NucView® 405 (bound to DNA): Ex/Em 429 nm/469 nm, NucView® 530 (bound to DNA): Ex/Em 528 nm/563 nm.

Certifications: NucView enzyme substrate technology is covered by U.S. Patent No. 8,092,784.
Ordering information: In addition to stand-alone NucView substrates, we offer a range of NucView® 488 live cell and apoptosis assay kits with detailed application protocols.
Customers Who Bought This Also Bought