ContactUsLinkComponent
FERROFLUID
Ward's® Chemistry Ferrofluid Demonstration
# de catalogue 470217-236
Fournisseur:  Ward's Science
FERROFLUID
Ward's® Chemistry Ferrofluid Demonstration
# de catalogue 470217-236
Fournisseur:  Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Ferrofluids Liquid Only Refill, 50 mL
  • No. of Lab Groups:
  • Cat. no.:
    470217-236
  • Includes:
  • Supplier No.:
    AD-FF0195-50ML
  • No. of Uses:
Spécifications
A propos de cet article
Amaze and Excite Your Students!
  • Ideal for teaching magnetism, forces and chemistry, Grades 9-12
  • Great demonstration for younger students, Grades 5-8
  • Learn about nanomaterials and their properties
  • Investigate technical applications for ferrofluids
Amaze and excite your students with ferrofluid, an incredible substance which is in liquid form until a strong magnetic field is brought near it. As the magnet comes closer to the ferrofluid, spikes begin to rise from it. Ask students to determine the reason for this peculiar reaction. Ferrofluid, a colloid with magnetic nanoparticles suspended in an oil base, was originally discovered by NASA scientists. Now ferrofluids are used to make rotating seals in high vacuum equipment and can be even found in high end speakers.
Ordering information: Materials include Vinyl Gloves, Jar with Lid, Instructions, Petri Dish, Ferrofluid, Glass Vials with Caps, Ceramic Block Magnet, Cylindrical Magnet, Nylon String, Plastic Jar with Lid, Modeling Clay, Test Tubes with Stoppers, and Bar Magnet. Pennies, Scotchgard™ spray, Water, Paper towels, Liquid dish soap, Lab coat/apron, Safety glasses are materials needed but not included.
Delivery: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Customers Who Bought This Also Bought