ELIMINASE®, Decontaminants, Decon Labs
ELIMINase, 250 ml, 12/case
Supplier: Decon Labs
Supplier #: 1101
Jump to:
Product Details & Documents
ELIMINase removes RNase, DNase, unwanted DNA and radioactive contamination.
- Phosphate free and non-toxic
- Safely disposable and totally rinsable for the decontaminated surface
- Eliminates the time-consuming method of autoclaving
It works instantly so there is no waiting. ELIMINase is an ideal substitute for using DiEthylPryoCarbonate (DEPC) which is a carcinogen. ELIMINase can be used on all surfaces including glass, plastic, metal and ceramic.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Type de conditionnement:12/case
- Désignation:ELIMINase
- Taille:250 ml