ELIMINASE®, Decontaminants, Decon Labs
ELIMINASE®, Decontaminants, Decon Labs
ELIMINase, 250 ml, 12/case
Catalog # 10798-804
Supplier:   Decon Labs
Supplier #:  1101
ELIMINASE®, Decontaminants, Decon Labs
Catalog # 10798-804
Supplier:  Decon Labs
CAS Number:  
ELIMINASE®, Decontaminants, Decon Labs
ELIMINASE®, Decontaminants, Decon Labs
ELIMINase, 250 ml, 12/case
CAS Number:  
Price
$43.60
Log in for availability
Each(1/Itm/Bx) Case of 12 (1/ Itm/Bx)
Product Details & Documents
ELIMINase removes RNase, DNase, unwanted DNA and radioactive contamination.
  • Phosphate free and non-toxic
  • Safely disposable and totally rinsable for the decontaminated surface
  • Eliminates the time-consuming method of autoclaving
It works instantly so there is no waiting. ELIMINase is an ideal substitute for using DiEthylPryoCarbonate (DEPC) which is a carcinogen. ELIMINase can be used on all surfaces including glass, plastic, metal and ceramic.
Documents
Specifications
Specifications
  • Type de conditionnement:
    12/case
  • Désignation:
    ELIMINase
  • Taille:
    250 ml