Order Entry
ContactUsLinkComponent
Test tube rack
Blue Epoxy-Coated Steel Wire Test Tube Racks
# de catalogue 470165-452
Fournisseur:  Wards
Test tube rack
Blue Epoxy-Coated Steel Wire Test Tube Racks
# de catalogue 470165-452
Fournisseur:  Wards

Spécifications

  • No. of Holes:
    24 (4×6)
  • Cat. no.:
    470165-452
  • For Tube O.D.:
    15 - 16 mm
  • Supplier No.:
    CH182015B

Spécifications

A propos de cet article

Coated steel wire racks are chemical and corrosion resistant and come in a variety of sizes.

  • Test tube rack featuring a sturdy epoxy-coated steel construction that will stand up to the wear and tear any lab
  • Withstands temperatures up to 121 °C (249.8 °F)

Coating provides a protective and smooth coat that is impervious to weak acids, bases, and salts.

Customers Who Bought This Also Bought