Order Entry
ContactUsLinkComponent
Clamp
Premium Three-Finger Clamp, Vinyl Coated
Catalog # 470165-396
Supplier:  Wards
Clamp
Premium Three-Finger Clamp, Vinyl Coated
Catalog # 470165-396
Supplier:  Wards

Specifications

  • Description:
    Premium Three-Finger Clamp with Bosshead, Vinyl Coated
  • Cat. no.:
    470165-396
  • Supplier No.:
    PRCMP01BH

Specifications

About this item

This premium lab clamp features thread-fitted actuating joints.

  • Maximum Clamp Opening: 8.5 cm
  • Connecting Rod: 14 cm
  • Powder-Coated Steel Construction
  • Vinyl-Coated Fingers
  • Thread-Fitted Actuating Joints

The clamp is made of powder-coated steel.

Thread-fitted actuating joints join the vinyl-coated fingers to a 10 mm connecting rod.

Customers Who Bought This Also Bought