Order Entry
Canada
ContactUsLinkComponent
Blue Epoxy-Coated Steel Wire Test Tube Racks
Catalogue # 470165-390
Fournisseur:  Wards
undefined
Blue Epoxy-Coated Steel Wire Test Tube Racks
Catalogue # 470165-390
Fournisseur:  Wards
Supplier Number:  CH182015A

Spécifications

  • No. of Holes:
    20 (4×5)
  • Cat. no.:
    470165-390
  • For Tube O.D.:
    18 - 20 mm
  • Supplier No.:
    CH182015A

Spécifications

À propos de cet article

Coated steel wire racks are chemical and corrosion resistant and come in a variety of sizes.

  • Test tube rack featuring a sturdy epoxy-coated steel construction that will stand up to the wear and tear any lab
  • Withstands temperatures up to 121 °C (249.8 °F)

Coating provides a protective and smooth coat that is impervious to weak acids, bases, and salts.

Frequently Bought Together