Vinyl GHS Labels "Pictogram",
# de catalogue 470165-352
Fournisseur: EMEDCO
Spécifications
- Description:Vinyl GHS picto labels
- Cat. no.:470165-352
- Packaging:39 labels/sheet
- Legend:Toxic
- Dimensions:15.9×16 mm (0.63×0.63")
- Supplier No.:GHMLSITOQ58
Spécifications
A propos de cet article
These labels help identify hazardous items in your facility.
- Labels stick to any surface
- OSHA Standard-Compliant
- Labels don't tear or peel
Die-cut GHS pictogram labels are pre-printed with the OSHA GHS required pictogram symbols. They have a white background with a red border and black graphics.
Ordering information: Each symbol is sold separately.
Customers Who Bought This Also Bought