Polypropylene Screw Caps, White, Wheaton
# de catalogue 10796-436
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap Size:24-400
- Cap Type:Solid Top
- Color:White
Spécifications
A propos de cet article
These caps are designed for use with WHEATON® glass bottles or vials.
- White surface for writing on cap
- Offered in a variety of liner materials
Polyethylene foam provides compressibility and sealing properties, and is excellent for use with environmental samples, pharmaceuticals, and diagnostic reagents. The Poly-vinyl liner provides an excellent moisture barrier, and is good for use with mild acids, alkalis, solvents, alcohols, oils, and aqueous products. The PTFE-faced foamed PE liner is ideal for use with high purity chemicals, strong acids, and solvents.
White polypropylene screw caps feature a clear surface for writing on cap.
Customers Who Bought This Also Bought