ContactUsLinkComponent
Ez-vision blue light DNA dye
VWR® EZ-Vision® Bluelight DNA Dye 10,000X
# de catalogue 10791-798
Fournisseur:  VWR
Numéro CAS:  
VWR® EZ-Vision® Bluelight DNA Dye 10,000X
# de catalogue 10791-798
Fournisseur:  VWR
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    EZ-Vision® Bluelight DNA dye 10,000X
  • Size:
    0.5 ml
  • Cat. no.:
    10791-798
  • Supplier No.:
    1B1680-0.5ML
Spécifications
A propos de cet article
EZ-Vision BlueLight is a sensitive, non mutagenic and environmentally safe fluorescent dye designed for gel staining.
  • In-gel and post-gel staining
  • Compatible with gel documentation using blue light excitation
  • Does not affect DNA integrity for downstream applications
Compatible either with a UV transilluminator, or a gel reader, equipped with blue light excitation such as a blue LED gel imaging system or a Dark Reader®.
EZ-Vision® Bluelight DNA dye, 10,000X, is a concentrated solution that can be diluted 10,000 times for use in either precast or post gel staining. One vial (0.5 mL) of 10,000X solution can be used to prepare at least 100 precast or post-stain 100 mini-gels.
Customers Who Bought This Also Bought