Accessories for Cimarec+™ Hotplates, Stirrers, and Stirring Hotplates, Thermo Scientific
# de catalogue 10791-516
Fournisseur: Thermo Fisher Scientific
Spécifications
- Description:PT1000 Hastelloy External Temperature Probe, 0–400°C
- Cat. no.:10791-516
- Supplier No.:CIC0001449