ContactUsLinkComponent
Cap
Screw Caps, 9 mm, Soft-Guard™, MicroSolv
# de catalogue 10790-742
Cap
Screw Caps, 9 mm, Soft-Guard™, MicroSolv
# de catalogue 10790-742
Spécifications
  • Cap Size:
    9-425 mm
  • Cap Type:
    Screw cap
  • Septum:
    Soft-Guard, White Silicone Rubber / Red PTFE, bonded
  • Material:
    PP
  • Cat. no.:
    10790-742
  • Cap Color:
    Blue
Spécifications
A propos de cet article
Screw caps for 12 x 32 autosampler vials.
Customers Who Bought This Also Bought