Screw Caps, 9 mm, Soft-Guard™, MicroSolv
# de catalogue 10790-742
Fournisseur: MicroSolv Technology Corp
Spécifications
- Cap Size:9-425 mm
- Cap Type:Screw cap
- Septum:Soft-Guard, White Silicone Rubber / Red PTFE, bonded
- Material:PP
- Cat. no.:10790-742
- Cap Color:Blue
Spécifications
A propos de cet article
Screw caps for 12 x 32 autosampler vials.
Customers Who Bought This Also Bought